Assay Details
| Gene Symbol: PGK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204985.2 | NP_990316.1 | 8 - 9 | 1028 | 85 | |
| GenBank mRNA | BX935600.2 | - | 9 - 10 | 865 | 85 | |
| L37101.1 | - | 8 - 9 | 1028 | 85 | ||
More Information
Set Membership: |
|

PGK2
Genomic Map