Assay Details
| Gene Symbol: AvBD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204993.1 | NP_990324.1 | 3 - 4 | 277 | 74 | |
| GenBank mRNA | AF033335.1 | - | 3 - 4 | 277 | 74 | |
| AF181951.1 | - | 2 - 3 | 189 | 74 | ||
| BX931239.1 | - | 3 - 4 | 285 | 74 | ||
More Information
Set Membership: |
|

AvBD1
Genomic Map