Assay Details
| Gene Symbol: RPL13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204999.1 | NP_990330.1 | 4 - 5 | 448 | 59 | |
| GenBank mRNA | D26318.1 | - | 4 - 5 | 448 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RPL13| Gene Symbol: RPL13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204999.1 | NP_990330.1 | 4 - 5 | 448 | 59 | |
| GenBank mRNA | D26318.1 | - | 4 - 5 | 448 | 59 | |
Set Membership: |
|