Assay Details
| Gene Symbol: HSPA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205003.2 | NP_990334.1 | 2 - 3 | 248 | 90 | |
| GenBank mRNA | AJ004940.1 | - | 2 - 3 | 249 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HSPA8| Gene Symbol: HSPA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205003.2 | NP_990334.1 | 2 - 3 | 248 | 90 | |
| GenBank mRNA | AJ004940.1 | - | 2 - 3 | 249 | 90 | |
Set Membership: |
|