Assay Details
| Gene Symbol: EFNB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205035.2 | NP_990366.1 | 2 - 3 | 653 | 63 | |
| XM_015278194.2 | XP_015133680.1 | 2 - 3 | 505 | 63 | ||
| XM_015278195.2 | XP_015133681.1 | 2 - 3 | 325 | 63 | ||
| GenBank mRNA | U72394.1 | - | 2 - 3 | 656 | 63 | |
More Information
Set Membership: |
|

EFNB1
Genomic Map