Assay Details
| Gene Symbol: PRPH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205038.1 | NP_990369.1 | 1 - 2 | 856 | 79 | |
| GenBank mRNA | AF031238.1 | - | 1 - 2 | 856 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRPH2| Gene Symbol: PRPH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205038.1 | NP_990369.1 | 1 - 2 | 856 | 79 | |
| GenBank mRNA | AF031238.1 | - | 1 - 2 | 856 | 79 | |
Set Membership: |
|