Assay Details
| Gene Symbol: DGCR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205040.1 | NP_990371.1 | 2 - 3 | 509 | 67 | |
| GenBank mRNA | AF048985.1 | - | 2 - 3 | 509 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DGCR6| Gene Symbol: DGCR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205040.1 | NP_990371.1 | 2 - 3 | 509 | 67 | |
| GenBank mRNA | AF048985.1 | - | 2 - 3 | 509 | 67 | |
Set Membership: |
|