Assay Details
| Gene Symbol: MGP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205044.2 | NP_990375.1 | 3 - 4 | 216 | 59 | |
| GenBank mRNA | Y13903.1 | - | 216 | 59 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MGP| Gene Symbol: MGP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205044.2 | NP_990375.1 | 3 - 4 | 216 | 59 | |
| GenBank mRNA | Y13903.1 | - | 216 | 59 | ||
Set Membership: |
|