Assay Details
| Gene Symbol: IBSP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205162.1 | NP_990493.1 | 2 - 3 | 176 | 76 | |
| GenBank mRNA | U10577.1 | - | 2 - 3 | 176 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

IBSP| Gene Symbol: IBSP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205162.1 | NP_990493.1 | 2 - 3 | 176 | 76 | |
| GenBank mRNA | U10577.1 | - | 2 - 3 | 176 | 76 | |
Set Membership: |
|