Assay Details
| Gene Symbol: HPCAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205234.1 | NP_990565.1 | 3 - 4 | 557 | 78 | |
| XM_015276024.1 | XP_015131510.1 | 2 - 3 | 509 | 78 | ||
| XM_015276026.2 | XP_015131512.1 | 2 - 3 | 598 | 78 | ||
| XM_015276027.2 | XP_015131513.1 | 2 - 3 | 590 | 78 | ||
| XM_015276028.1 | XP_015131514.1 | 2 - 3 | 663 | 78 | ||
| XM_025148553.1 | XP_025004321.1 | 3 - 4 | 3169 | 78 | ||
| XM_025148554.1 | XP_025004322.1 | 2 - 3 | 486 | 78 | ||
| XM_025148555.1 | XP_025004323.1 | 2 - 3 | 504 | 78 | ||
| GenBank mRNA | X80875.1 | - | 557 | 78 | ||
More Information
Set Membership: |
|

HPCAL1
Genomic Map