Assay Details
| Gene Symbol: CD8BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205247.2 | NP_990578.2 | 4 - 5 | 983 | 90 | |
| GenBank mRNA | Z26484.1 | - | 983 | 90 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CD8BP| Gene Symbol: CD8BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205247.2 | NP_990578.2 | 4 - 5 | 983 | 90 | |
| GenBank mRNA | Z26484.1 | - | 983 | 90 | ||
Set Membership: |
|