Assay Details
| Gene Symbol: ITGB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205251.1 | NP_990582.1 | 4 - 5 | 439 | 65 | |
| XM_015289134.2 | XP_015144620.1 | 5 - 6 | 833 | 65 | ||
| XM_015289135.2 | XP_015144621.1 | 5 - 6 | 832 | 65 | ||
| XM_015289136.2 | XP_015144622.1 | 4 - 5 | 706 | 65 | ||
| GenBank mRNA | X71786.1 | - | 439 | 65 | ||
More Information
Set Membership: |
|

ITGB2
Genomic Map