Assay Details
| Gene Symbol: GCG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190165.4 | NP_001177094.1 | 3 - 4 | 362 | 92 | |
| NM_205260.5 | NP_990591.1 | 3 - 4 | 362 | 92 | ||
| NR_073182.3 | 3 - 4 | 351 | 92 | |||
| XM_015289639.2 | XP_015145125.1 | 4 - 5 | 4133 | 92 | ||
| GenBank mRNA | DQ185930.1 | - | 3 - 4 | 362 | 92 | |
| DQ185931.1 | - | 3 - 4 | 362 | 92 | ||
| DR429295.1 | - | 3 - 4 | 358 | 92 | ||
| S78477.1 | - | 3 - 4 | 360 | 92 | ||
| Y07539.1 | - | 352 | 92 | |||
More Information
Set Membership: |
|

GCG
Genomic Map