Assay Details
| Gene Symbol: CCN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205268.1 | NP_990599.1 | 4 - 5 | 778 | 115 | |
| GenBank mRNA | X59284.1 | - | 778 | 115 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCN3| Gene Symbol: CCN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205268.1 | NP_990599.1 | 4 - 5 | 778 | 115 | |
| GenBank mRNA | X59284.1 | - | 778 | 115 | ||
Set Membership: |
|