Assay Details
| Gene Symbol: RXRG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205294.1 | NP_990625.1 | 9 - 10 | 1313 | 64 | |
| XM_015290246.2 | XP_015145732.1 | 9 - 10 | 1422 | 64 | ||
| XM_015290248.2 | XP_015145734.1 | 9 - 10 | 1602 | 64 | ||
| XM_015290249.2 | XP_015145735.1 | 9 - 10 | 1402 | 64 | ||
| GenBank mRNA | X58997.1 | - | 1313 | 64 | ||
More Information
Set Membership: |
|

RXRG
Genomic Map