Assay Details
| Gene Symbol: YES1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205301.1 | NP_990632.1 | 4 - 5 | 590 | 63 | |
| GenBank mRNA | X12461.1 | - | 590 | 63 | ||
| X13207.1 | - | 486 | 63 | |||
More Information
Set Membership: |
|

YES1
Genomic Map