Assay Details
| Gene Symbol: ETS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205312.1 | NP_990643.1 | 4 - 5 | 574 | 67 | |
| GenBank mRNA | X07202.1 | - | 574 | 67 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ETS2| Gene Symbol: ETS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205312.1 | NP_990643.1 | 4 - 5 | 574 | 67 | |
| GenBank mRNA | X07202.1 | - | 574 | 67 | ||
Set Membership: |
|