Assay Details
| Gene Symbol: CDK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205314.1 | NP_990645.1 | 4 - 5 | 412 | 92 | |
| XM_015288063.2 | XP_015143549.1 | 4 - 5 | 466 | 92 | ||
| XM_015288064.2 | XP_015143550.1 | 4 - 5 | 502 | 92 | ||
| GenBank mRNA | X16881.1 | - | 412 | 92 | ||
More Information
Set Membership: |
|

CDK1
Genomic Map