Assay Details
| Gene Symbol: CALB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205316.1 | NP_990647.1 | 2 - 3 | 165 | 144 | |
| GenBank mRNA | BX950496.1 | - | 2 - 3 | 163 | 144 | |
| X62866.1 | - | 165 | 144 | |||
| Y00625.1 | - | 150 | 144 | |||
More Information
Set Membership: |
|

CALB2
Genomic Map