Assay Details
| Gene Symbol: CA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205317.1 | NP_990648.1 | 6 - 7 | 880 | 71 | |
| GenBank mRNA | X04810.1 | - | 656 | 71 | ||
| Z14957.1 | - | 880 | 71 | |||
More Information
Set Membership: |
|

CA2
Genomic Map