Assay Details
| Gene Symbol: RPLP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205322.1 | NP_990653.1 | 1 - 2 | 150 | 63 | |
| GenBank mRNA | CR353684.1 | - | 1 - 2 | 150 | 63 | |
| X13876.1 | - | 150 | 63 | |||
More Information
Set Membership: |
|

RPLP1
Genomic Map