Assay Details
| Gene Symbol: RPL27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205337.1 | NP_990668.1 | 3 - 4 | 276 | 124 | |
| GenBank mRNA | BX930617.1 | - | 2 - 3 | 374 | 124 | |
| CR524112.1 | - | 2 - 3 | 517 | 124 | ||
| X56852.1 | - | 276 | 124 | |||
More Information
Set Membership: |
|

RPL27
Genomic Map