Assay Details
| Gene Symbol: RPL27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205337.1 | NP_990668.1 | 4 - 5 | 391 | 71 | |
| GenBank mRNA | X56852.1 | - | 391 | 71 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RPL27| Gene Symbol: RPL27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205337.1 | NP_990668.1 | 4 - 5 | 391 | 71 | |
| GenBank mRNA | X56852.1 | - | 391 | 71 | ||
Set Membership: |
|