Assay Details
| Gene Symbol: GABRB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205346.3 | NP_990677.1 | 6 - 7 | 762 | 113 | |
| GenBank mRNA | X54243.1 | - | 775 | 113 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GABRB3| Gene Symbol: GABRB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205346.3 | NP_990677.1 | 6 - 7 | 762 | 113 | |
| GenBank mRNA | X54243.1 | - | 775 | 113 | ||
Set Membership: |
|