Assay Details
| Gene Symbol: MAP2K2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205388.1 | NP_990719.1 | 9 - 10 | 1052 | 83 | |
| XM_015299918.2 | XP_015155404.1 | 9 - 10 | 1081 | 83 | ||
| GenBank mRNA | L28703.1 | - | 9 - 10 | 1052 | 83 | |
More Information
Set Membership: |
|

MAP2K2
Genomic Map