Assay Details
| Gene Symbol: SLC2A14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205511.1 | NP_990842.1 | 7 - 8 | 1053 | 68 | |
| GenBank mRNA | M37785.1 | - | 7 - 8 | 1053 | 68 | |
More Information
Set Membership: |
|

SLC2A14
Genomic Map