Assay Details
| Gene Symbol: CCT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_206878.1 | NP_996761.1 | 1 - 2 | 248 | 66 | |
| GenBank mRNA | AJ251587.1 | - | 1 - 2 | 248 | 66 | |
| AJ719421.1 | - | 1 - 2 | 245 | 66 | ||
| BX932477.1 | - | 1 - 2 | 239 | 66 | ||
More Information
Set Membership: |
|

CCT4
Genomic Map