Assay Details
| Gene Symbol: LGALS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001302800.1 | NP_001289729.1 | 5 - 6 | 612 | 91 | |
| XM_015287256.2 | XP_015142742.1 | 5 - 6 | 558 | 91 | ||
| GenBank mRNA | AF479564.1 | - | 2 - 3 | 904 | 91 | |
| AF479565.1 | - | 1 - 2 | 518 | 91 | ||
| BX261386.4 | - | 5 - 6 | 564 | 91 | ||
| CD738329.1 | - | 3 - 4 | 318 | 91 | ||
| EF429082.1 | - | 4 - 5 | 569 | 91 | ||
| U50339.1 | - | 4 - 5 | 1027 | 91 | ||
More Information
Set Membership: |
|

LGALS3
Genomic Map