Assay Details
| Gene Symbol: CHST10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030343.1 | NP_001025514.1 | 2 - 3 | 187 | 77 | |
| XM_015277674.2 | XP_015133160.1 | 2 - 3 | 1945 | 77 | ||
| XM_025153288.1 | XP_025009056.1 | 4 - 5 | 2358 | 77 | ||
| XM_025153302.1 | XP_025009070.1 | 3 - 4 | 2266 | 77 | ||
| XM_025153319.1 | XP_025009087.1 | 2 - 3 | 259 | 77 | ||
| GenBank mRNA | AJ720840.1 | - | 2 - 3 | 187 | 77 | |
More Information
Set Membership: |
|

CHST10
Genomic Map