Assay Details
| Gene Symbol: HEXA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030390.1 | NP_001025561.1 | 11 - 12 | 1336 | 59 | |
| GenBank mRNA | AJ720938.1 | - | 11 - 12 | 1336 | 59 | |
More Information
Set Membership: |
|

HEXA
Genomic Map