Assay Details
| Gene Symbol: CRABP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030539.1 | NP_001025710.1 | 2 - 3 | 383 | 103 | |
| GenBank mRNA | AB124570.1 | - | 2 - 3 | 383 | 103 | |
| EU334504.1 | - | 2 - 3 | 291 | 103 | ||
| X53701.1 | - | 223 | 103 | |||
More Information
Set Membership: |
|

CRABP1
Genomic Map