Assay Details
| Gene Symbol: NRG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030544.1 | NP_001025715.1 | 5 - 6 | 499 | 90 | |
| XM_015278876.2 | XP_015134362.1 | 5 - 6 | 534 | 90 | ||
| XM_015278877.2 | XP_015134363.1 | 5 - 6 | 534 | 90 | ||
| GenBank mRNA | AJ720374.1 | - | 5 - 6 | 499 | 90 | |
More Information
Set Membership: |
|

NRG4
Genomic Map