Assay Details
| Gene Symbol: ITGB1BP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030550.1 | NP_001025721.1 | 3 - 4 | 418 | 96 | |
| GenBank mRNA | AJ719838.1 | - | 3 - 4 | 418 | 96 | |
| BX935109.2 | - | 514 | 96 | |||
| BX935895.2 | - | 3 - 4 | 412 | 96 | ||
| CR352634.1 | - | 3 - 4 | 411 | 96 | ||
More Information
Set Membership: |
|

ITGB1BP3
Genomic Map