Assay Details
| Gene Symbol: FAM96A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030555.1 | NP_001025726.1 | 1 - 2 | 157 | 97 | |
| XM_025153863.1 | XP_025009631.1 | 1 - 2 | 177 | 97 | ||
| XM_025153864.1 | XP_025009632.1 | 1 - 2 | 260 | 97 | ||
| GenBank mRNA | AJ720392.1 | - | 1 - 2 | 157 | 97 | |
More Information
Set Membership: |
|

FAM96A
Genomic Map