Assay Details
| Gene Symbol: GALK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030557.1 | NP_001025728.1 | 9 - 10 | 1246 | 95 | |
| GenBank mRNA | AJ719347.1 | - | 9 - 10 | 1246 | 95 | |
More Information
Set Membership: |
|

GALK2
Genomic Map