Assay Details
| Gene Symbol: NUTF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030562.2 | NP_001025733.2 | 4 - 5 | 301 | 71 | |
| NM_001305547.1 | NP_001292476.1 | 3 - 4 | 262 | 71 | ||
| NM_001305548.1 | NP_001292477.1 | 3 - 4 | 397 | 71 | ||
| XM_015279126.2 | XP_015134612.1 | 3 - 4 | 253 | 71 | ||
| GenBank mRNA | AJ719600.1 | - | 4 - 5 | 301 | 72 | |
| BU116275.1 | - | 3 - 4 | 262 | 71 | ||
| BU144410.1 | - | 3 - 4 | 248 | 71 | ||
| BU345205.1 | - | 3 - 4 | 397 | 71 | ||
More Information
Set Membership: |
|

NUTF2
Genomic Map