Assay Details
| Gene Symbol: DYNC1LI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030573.1 | NP_001025744.1 | 9 - 10 | 1290 | 67 | |
| XM_015292500.2 | XP_015147986.2 | 9 - 10 | 2612 | 67 | ||
| GenBank mRNA | AJ719330.1 | - | 9 - 10 | 1290 | 67 | |
More Information
Set Membership: |
|

DYNC1LI2
Genomic Map