Assay Details
| Gene Symbol: TLDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030575.1 | NP_001025746.1 | 4 - 5 | 597 | 74 | |
| XM_015292554.2 | XP_015148040.2 | 4 - 5 | 977 | 74 | ||
| GenBank mRNA | AJ720309.1 | - | 4 - 5 | 597 | 74 | |
More Information
Set Membership: |
|

TLDC1
Genomic Map