Assay Details
| Gene Symbol: CBFA2T3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030580.1 | NP_001025751.1 | 3 - 4 | 765 | 100 | |
| XM_025154189.1 | XP_025009957.1 | 4 - 5 | 1668 | 100 | ||
| XM_025154190.1 | XP_025009958.1 | 4 - 5 | 1672 | 100 | ||
| XM_025154191.1 | XP_025009959.1 | 4 - 5 | 883 | 100 | ||
| XM_025154192.1 | XP_025009960.1 | 4 - 5 | 955 | 100 | ||
| XM_025154193.1 | XP_025009961.1 | 3 - 4 | 805 | 100 | ||
| GenBank mRNA | AJ851739.1 | - | 3 - 4 | 765 | 100 | |
| AY714074.1 | - | 4 - 5 | 575 | 100 | ||
| AY714075.1 | - | 3 - 4 | 451 | 100 | ||
More Information
Set Membership: |
|

CBFA2T3
Genomic Map