Assay Details
| Gene Symbol: PFKFB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001361249.1 | NP_001348178.1 | 657 | 65 | ||
| XM_025146276.1 | XP_025002044.1 | 651 | 65 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PFKFB1| Gene Symbol: PFKFB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001361249.1 | NP_001348178.1 | 657 | 65 | ||
| XM_025146276.1 | XP_025002044.1 | 651 | 65 | |||
Set Membership: |
|