Assay Details
| Gene Symbol: BAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030590.1 | NP_001025761.1 | 1 - 2 | 380 | 54 | |
| GenBank mRNA | AJ851614.1 | - | 1 - 2 | 380 | 54 | |
More Information
Set Membership: |
|

BAP1
Genomic Map