Assay Details
| Gene Symbol: RPS14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030619.1 | NP_001025790.1 | 3 - 4 | 353 | 91 | |
| XM_015293784.2 | XP_015149270.1 | 3 - 4 | 639 | 91 | ||
| XM_015293785.2 | XP_015149271.1 | 3 - 4 | 550 | 91 | ||
| GenBank mRNA | AJ721016.1 | - | 3 - 4 | 353 | 91 | |
More Information
Set Membership: |
|

RPS14
Genomic Map