Assay Details
| Gene Symbol: RPS14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030619.1 | NP_001025790.1 | 4 - 5 | 429 | 75 | |
| XM_015293784.2 | XP_015149270.1 | 4 - 5 | 715 | 75 | ||
| XM_015293785.2 | XP_015149271.1 | 4 - 5 | 626 | 75 | ||
| GenBank mRNA | AJ721016.1 | - | 4 - 5 | 429 | 75 | |
More Information
Set Membership: |
|

RPS14
Genomic Map