Assay Details
| Gene Symbol: ATP6AP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030801.2 | NP_001025972.2 | 4 - 5 | 498 | 68 | |
| XM_015301559.2 | XP_015157045.1 | 4 - 5 | 485 | 68 | ||
| GenBank mRNA | AB188299.2 | - | 4 - 5 | 498 | 68 | |
| AJ719432.1 | - | 4 - 5 | 463 | 68 | ||
| CR406923.1 | - | 2 - 3 | 152 | 68 | ||
More Information
Set Membership: |
|

ATP6AP2
Genomic Map