Assay Details
| Gene Symbol: WBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030824.1 | NP_001025995.1 | 4 - 5 | 311 | 77 | |
| XM_015275479.2 | XP_015130965.1 | 3 - 4 | 305 | 77 | ||
| XM_015275483.2 | XP_015130969.1 | 2 - 3 | 247 | 77 | ||
| GenBank mRNA | AJ851443.1 | - | 4 - 5 | 311 | 77 | |
More Information
Set Membership: |
|

WBP4
Genomic Map