Assay Details
| Gene Symbol: UFM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030827.1 | NP_001025998.1 | 5 - 6 | 225 | 95 | |
| XM_025150475.1 | XP_025006243.1 | 4 - 5 | 390 | 95 | ||
| GenBank mRNA | AJ719377.1 | - | 5 - 6 | 225 | 95 | |
| CR387688.1 | - | 4 - 5 | 270 | 95 | ||
More Information
Set Membership: |
|

UFM1
Genomic Map