Assay Details
| Gene Symbol: TAF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030841.2 | NP_001026012.1 | 6 - 7 | 2780 | 63 | |
| XM_015293603.2 | XP_015149089.1 | 5 - 6 | 9870 | 63 | ||
| GenBank mRNA | AJ851411.1 | - | 6 - 7 | 2783 | 63 | |
More Information
Set Membership: |
|

TAF3
Genomic Map