Assay Details
| Gene Symbol: NFS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030847.1 | NP_001026018.1 | 7 - 8 | 811 | 64 | |
| GenBank mRNA | AJ720039.1 | - | 7 - 8 | 811 | 64 | |
More Information
Set Membership: |
|

NFS1
Genomic Map