Assay Details
| Gene Symbol: TAF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030923.1 | NP_001026094.1 | 423 | 63 | ||
| XM_015298954.2 | XP_015154440.1 | 386 | 63 | |||
| XM_015298955.2 | XP_015154441.1 | 782 | 63 | |||
| XM_025143708.1 | XP_024999476.1 | 573 | 63 | |||
| GenBank mRNA | AJ719313.1 | - | 423 | 63 | ||
More Information
Set Membership: |
|

TAF8
Genomic Map