Assay Details
| Gene Symbol: PCMTD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031019.1 | NP_001026190.1 | 4 - 5 | 722 | 112 | |
| XM_015282593.2 | XP_015138079.1 | 4 - 5 | 989 | 112 | ||
| XM_015282594.2 | XP_015138080.1 | 4 - 5 | 777 | 112 | ||
| XM_015282595.2 | XP_015138081.1 | 3 - 4 | 444 | 112 | ||
| GenBank mRNA | AJ719321.1 | - | 4 - 5 | 722 | 112 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015282595.2 transcripts.
Set Membership: |
|

PCMTD1
Genomic Map